PYREX® Burets, Class B, Straight Bore Fluoropolymer Resin Stopcock, Colored Scale, Corning
Proveedor: Corning
PYREX®
2122A-10 2122A-100 2122A-25 2122A-50
17452-120CS 2100 MXN
17452-164 17452-120 17452-142 17452-109
PYREX® Burets, Class B, Straight Bore Fluoropolymer Resin Stopcock, Colored Scale, Corning
Buretas
Borosilicate glass.
  • Microfinish of the barrel provides a precision fit
  • Durable colored markings
These burets are suitable for use in school and institutional laboratories where the accuracy of class A models is not required.
The tips are carefully drawn for accuracy to insure proper drainage rate.
Información de suministro: Burets are supplied with a 2 mm PTFE stopcock plug and dust cover.