Basic™ 47 and Basic 70 Laboratory Hoods, Labconco®
Proveedor: Labconco
Basic™
2247500 2246701 2246703 2246300 2247501 2246605 2246704 2246600 2246501 2246400 2247300 2246604 2246601 2247400 2246301 2246500 2246705 2246606 2246603 2246602 2246700 2246706 2246702 2246401 2247401 2247301
30152-518EA 144555.84 MXN
30152-562 30152-518 30140-230 30140-232 30140-210 30140-234 30140-236 30140-214 30140-238 30140-216 30140-218 30152-612 30152-610 30140-240 30140-242 30140-220 30140-200 30140-222 30140-244 30140-224 30140-202 30140-204 30140-226 30140-228 30140-206 30140-208
Basic™ 47 and Basic 70 Laboratory Hoods, Labconco®
Vitrinas de gases y recintos Campanas de gases químicos
The Basic™ hood is ideal for rooms with ceilings below 2.4 m, 8" and for shallow countertops.
  • Blowers Available
  • Safety Glass
  • Steel Construction
  • Vapor-Proof, Incandescent Light Fixture
The hood is furnished with a 0.5 cm (3/16") thick safety glass and a 25.4 cm (10") dia
The hood is available with or without a blower, which can be mounted in or above the hood or on the roof. A 1/3 hp motor is included with Basic 47 hoods that have a blower (except 30152-610 and 612, which have a 1/2 hp motor). Basic 70 hoods with a blower have a 3/4 hp motor.
duct collar. The interior and exterior are constructed of glacier-white, epoxy-coated, 16-gauge steel. The hood includes a removable front panel, a removable air foil, a two-piece adjustable baffle, and a vapor-proof, incandescent light fixture. The right-side fixture panel is factory prepared to accept up to three service fixtures, one duplex electrical receptacle, and a Guardian air monitor. The right, interior side wall is factory prepared for one wall-mounted cup sink with a turret, and the hood liner top is constructed for two customer-supplied apparatus rods.
Certificaciones: The 115V models are ETL and ETL-C listed; 230V models are CE marked.
Información para pedidos: A work surface and base cabinet or stand are required for operation (all sold separately). A service fixture, receptacle, Guardian monitor, cup sink, and turret kits are also available. For unassembled Basic hoods, please contact your Avantor sales representative. Switches, electrical wiring, and connections are not included with explosion-proof units. Wiring and switches must be added after installation. Field-installed electrical wiring of explosion-proof units is not provided.