MetroMax Q™ Heavy-Duty Shelves and Posts, Metro™
Proveedor: Metro International
MetroMax Q® MetroMax Q™
MQ2448G MQ27UPE MQ63UPE Q2148SM MQ1854G MQ2160G MQ27PE MQ2142G MQ1830G MQ1842G Q2172SM Q2172SM MQ74UPE MQ2130G MQ2154G MQ2124G MQ2472G MQ1872G MQ1848G MQ2430G MQ2136G MQ86PE Q2160SM MQ2172G MQ1824G MQ54PE MQ13UPE MQ33UPE MQ2460G MQ13PE MQ2442G MQ2454G MQ86UPE MQ54UPE MQ33PE Q2136SM Q2136SM MQ74PE Q2160SM MQ2424G MQ1860G MQ2436G Q2148SM
82003-270EA 2991.18 MXN
83007-468 83007-446 83007-448 22233-504 83007-464 83007-444 83007-466 CA22233-478 22233-482 82003-268 22233-480 22233-484 82003-274 76240-550 82003-278 82003-276 83007-470 83007-472 83007-450 82003-270 83007-478 83007-456 22233-478 83007-458 22233-498 83007-452 83007-474 83007-454 83007-476 CA22233-480 CA22233-484 22233-496 CA22233-482 22233-494 82003-284 82003-282 82003-266 82003-264 83007-482 83007-460 83007-462 82003-280 83007-480
MetroMax Q™ Heavy-Duty Shelves and Posts, Metro™
Mobiliario Estantes
Maximize the storage space with this heavy-duty shelving.
  • Shelves are made of steel with electroplated substrate and highly durable, abrasion-resistant epoxy finish
  • Corner Lock Release™ allows adjustment of polymer shelves in seconds without tools
  • Open-grid construction allows air circulation and reduces dust and moisture build-up
MetroMax Q™ products are recommended for continuous use in temperatures from –29 to 52 °C (–20 to 125 °F), with acceptable intermittent exposure to 93°C (200°F). MetroMax Q™ shelves can quickly become mobile by removing the threaded leveling feet from the mobile posts and adding MetroMax iQ™ stem casters (see 19766-254 series).
Shelves up to 121.9cm (48") in length support up to 362.9kg (800lbs.) per level. Shelves between 121.9cm (48") and 182.9cm (72") in length can support up to 272.2kg (600lbs.) per level. The included gridded polymer shelf mats lift off for easy cleaning. Shelf mats feature an antimicrobial coating to protect against bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation. Smooth, rounded epoxy-coated posts are adjustable at 2.5cm (1") increments and are fitted with adjustable leveling bolts to compensate for uneven surfaces. Optional solid mat overlay kits turn standard grid shelves into solid shelves for spill containment.
Información para pedidos: Shelves feature a manufacturer's 15-year warranty against rust; grid shelf mats feature a lifetime warranty. Order shelves, posts, and solid mat overlay kits separately. Shelves come with frame, grid shelf mat, and wedge connectors.