Silica Sorbents and Bonded Silica Sorbents for Flash and Column Chromatography (60Å Pore Size), Agela Technologies
Proveedor: Agela Technologies
CS605025-P CS605001-G CS605001-P CS605025-G CS608025-P CS608005-P CS605005-P CS605005-G CS608025-G
97006-674 97006-662 97006-660 97006-670 97006-658 97006-668 97006-656 97006-678 97006-676
Silica Sorbents and Bonded Silica Sorbents for Flash and Column Chromatography (60Å Pore Size), Agela Technologies
Sorbentes para cromatografía Sorbentes para SPE
Bulk silica sorbents and bonded silica sorbents for flash and column chromatography applications.