PYREX® Iodine Determination Flasks, [ST] Stopper, Corning
Proveedor: Corning
PYREX®
5400-250 5400-500 5400-125
89090-780CS 3544.64 MXN
89090-780 89091-410 89090-640
PYREX® Iodine Determination Flasks, [ST] Stopper, Corning
Matraces Frascos para yodo
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Blown in a specially-designed mold to ensure uniformity and proper contour. The stopper handle projects above the liquid seal trough to facilitate removal of the stopper. With No. 22 [ST] stopper. ASTM D29 and D555.