Rhino-Pro™ Nasal Cytology Curettes, ASI
Proveedor: Arlington Scientific
Rhino-probe™
SY-96-0905 SY-96-0925 SY-96-0905 SY-96-0925
80087-272EA 11491.42 MXN
80087-272 CA80087-274 CA80087-272 80087-274
Rhino-Pro™ Nasal Cytology Curettes, ASI
Curetas
Plastic disposable nasal curettes allow the acquisition of a large number of intact cells to interpret and diagnose numerous nasal and respiratory disorders.
The curettes permit more accurate diagnosis of chronic rhinitis and aid in the detection of respiratory syncytial virus in children. They cause minimal to no discomfort and provide consistent and uniform mucosal samples with 100–1000 times more cells than any other method. Cells do not adhere to the curettes and no degranulation or distortion is seen after staining. Curettes also allow screening for infectious disease. Sterile.