SP Bel-Art Acetal Plastic Tubing Clamps, Autoclavable, Bel-Art Products, a part of SP
Proveedor: Bel-Art
F18228-0000 F18229-0000 F18227-0000
63022-405CS 4281.08 MXN
63022-405 63022-403 63022-407
SP Bel-Art Acetal Plastic Tubing Clamps, Autoclavable, Bel-Art Products, a part of SP
Abrazaderas Pinzas para tubos
These acetal clamps will not distort, rust, or corrode.
  • Acetal plastic construction
  • Multiple models available with varying position options
  • Multi-position clamps provide full range of control over closure
Single Position Mini Clamp: Single-position, on-off clamp for use with thin-walled tubing under 4.7 mm O.D
12-Position Maxi Clamp: 12-position ratchet control from full open to complete closure. For tubing up to 19 mm O.D. (3/4").
15-Position Mid-Range Clamp: 15-position stepped control throttles flow from full open to complete closure. Fits tubing from 3.2–11 mm O.D. (1/87/16").
(3/16"). Gives positive shut-off with finger tip control.