Day Pinchcock Clamp
4433
21730-001PK 335.16 MXN
21730-001
Day Pinchcock Clamp
For rubber tubing
Heavy stainless steel wire. Self-closing, quick acting. Can be clamped on tubing at any point without disconnecting the apparatus. For tubing up to 9.5mm (3/8") outside diameter.