Corning® Polypropylene Snap-Seal Sample Containers, Corning
Proveedor: Corning
Corning®
1730-8 1730-4H 1730-10 1730-4L 1730-5X 1730-2C
16126-018CS 1842.81 MXN
16126-018 16126-010 16126-022 16126-030 16126-014 16126-026
Corning® Polypropylene Snap-Seal Sample Containers, Corning
Recipientes de muestras Contenedores, uso general
PP, disposable, translucent.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both millilitres and ounces
  • Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.