VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
Proveedor: VWR
VWR®
K2600-VWR K2502-VWR K2508-VWR K2511-VWR
15070-290CS 3856.22 MXN
15070-296 15070-294 15070-292 15070-290
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
Recipientes de muestras Vasos de muestras para analizadores
These transparent polystyrene sample cups are resistant to most dilute aqueous solutions.
  • Compatible with most autoanalyzers
  • Wide base prevents spillage
Cups are available for use with Technicon®, Abbott®, Hitachi®, and most other autoanalyzers
Autoanalyzer sample cups are not autoclavable.
The wide-base design prevents tipping and spilling. The conical bottom is standard and recommended for microsampling.
Información para pedidos: The 2.0 mL cup (15070-290) also fits the Hyland laser-beam analyzer. Push-on polyethylene cap (15070-089) fits cups 15070-290, -293, and -294.