Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Proveedor: Starplex Scientific
Sanicult™
B30U-S5 B30U-DW B30U-S B30U-S5 B30U-S
15001-168CS 5411.82 MXN
15001-168 CA11006-522 CA73250-020 14216-280 CA73250-017
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Sistemas para monitorización Control higiene
Swab test kits are for hard-to-reach places, surfaces, viscous liquids, semi-solids, and powders.
Ideal for applications in food and beverage, engineering, and petrochemical industries.
The kit consists of a sterile swab and buffered solution. To use, remove swab from diluent vial, swab sample area, return swab to vial, and mix well. The sample is easily stored and readily transported. Choose from 5 ml or 8.8 ml diluent solution volumes.