VWR® Thermometer-Hygrometer-Barometer
Proveedor: VWR
VWR®
61502-0211
89030-410EA 2246.64 MXN
89030-410
VWR® Thermometer-Hygrometer-Barometer
Higrómetros Higrómetros analógicos
Triple scale barometers feature readings in cm Hg, in
  • Stainless steel and brass construction
  • Can be recalibrated easily
  • Feature single-scale thermometer and humidity
Complete with hole for wall mounting.
Hg and mbar.