Syringe Pumps, KD Scientific
780232 780230 780200 780220 780210 780212
52459-004EA 97949.87 MXN
52459-016 52459-004 52459-024 52459-008 52459-028 52459-012
Syringe Pumps, KD Scientific
These microprocessor-controlled syringe pumps from KD Scientific are designed for applications requiring low, pulse-free flow rates
  • Simple menu-driven setup to program syringe diameter, flowrate and dispense volume
  • Backlit display, automatic shut-off, settings can be reviewed and changed during operation, last settings are stored in memory for repeatability
  • Multiple syringe pumps
  • Capacity 10 µL to 140 ml up to 10 syringes (depending on syringe size)
  • Built-in RS-232-C and TTL interfaces for easy external control
  • Hold glass or plastic syringes
  • Available as infusion and infusion/withdrawal models
The dispensed volume is continuously displayed and when the preset volume has been dispensed, the pump shuts off automatically.
Model 200 is a dual-syringe pump that has a broad speed range and holds a wide range of syringe sizes. It combines all of the features of the single-syringe pump but includes an optical encoder stall detection; built-in RS-232C interface for connecting up to 100 units; TTL interface for foot switch, computer, outputs for run indicator and valve control; and the ability to review or change settings during operation.
Model 210 is another dual-syringe pump, combining all of the features of the Model 200 with five operation modes, and independent rate and volume settings for both infusion and withdrawal.
Model 220 and Model 230 are ideal for applications requiring multiple syringes. Model 220 is based on the 200, and Model 230 on the 210, but both have been modified to accept up to ten syringes.
Infusion/withdrawal pumps 210P and 230P include an added keypad programmable option.
Información para pedidos: Models 200, 210, 220, and 230 have a liner force of 18 kg (40 lbs.), 0.165 micron (1/16 step) Advance Per Microstep, a Maximum Step Rate of 1600 steps/sec. (1/2 Step), and a Minimum Step Rate of 1 step/30 sec. For an optional keypad programmable feature for Models 200 and 220, contact your VWR sales representative. Pumps with a multiple syringe capacity can hold four with volumes of 100–140mL, six with volumes of 20–60mL, and ten with volumes of 10µL–10mL.
Precaución: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.