Esta buscando: Placas de cultivo
31  results were found
(IVA no incluido)
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
SearchResultCount:"31"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: VWR International
Descripción: Clearly superior, high-quality Petri dishes are manufactured from virgin crystal grade polystyrene for optimum flatness and high optical clarity.
Proveedor: VWR International
Descripción: These Petri dishes are suitable for use in pharmaceuticals, food processing, hospital environment and other industries for samples purposes.
Proveedor: VWR International
Descripción: These slippable polystyrene Petri dishes provide optimal flatness.
Proveedor: Corning
Descripción: Dishes are constructed with crystal-grade polystyrene and a flat bottom surface for distortion-free optics
Numero del catalogo: (25388-640)
Proveedor: Cytiva
Descripción: PS dish, cellulose pad. Ideal for microbiological analysis when performing the membrane filter technique. Petri dishes with absorbent pads can be used...
Numero del catalogo: (90000-004)
Proveedor: BECTON DICKINSON DE MEXICO, S. MX
Descripción: RODAC® (Replicate Organism Detection and Counting) was developed for environmental studies to determine levels of microbial contamination of surfaces
Numero del catalogo: (28145-473)
Proveedor: Cytiva
Descripción: The Analyslide® Petri dish has a rectangular base and a circular chamber that accepts a 47 mm membrane filter. The sample is protected during mic...
Numero del catalogo: (73521-006)
Proveedor: Thermo Fisher Scientific
Descripción: These polystyrene dishes with flat-bottom wells incorporate the Nunclon™Δ treated surface for optimal cell attachment and growth
Numero del catalogo: (75801-452)
Proveedor: Corning
Descripción: The Corning® Gosselin™ Round Petri Dish is manufactured from crystal polystyrene, and designed for use in microbiology testing applications
Numero del catalogo: (76331-200)
Proveedor: Thermo Fisher Scientific
Descripción: Enhance cell attachment to the culture surface with the extracellular matrix coating on Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes.
Proveedor: Corning
Descripción: Flat, clear petri dishes are manufactured from soda lime silica glass and will withstand repeated sterilization (wet or dry)
Numero del catalogo: (25382-334)
Proveedor: Thermo Fisher Scientific
Descripción: Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs with Thermo Scientific™ Nunc™ Cell ...
Proveedor: Corning
Descripción: Optically clear PS. Available with a choice of treated surface. Corning® CellBIND® surface increases surface wetability for more even and consistent c...
Numero del catalogo: (470202-134)
Proveedor: CORNING CA
Descripción: Flat, clear dishes that can withstand repeated sterilization (wet or dry).
Numero del catalogo: (73521-368)
Proveedor: Thermo Fisher Scientific
Descripción: Round, Vented, I.D. 136 mm, Area 145 cm2
Numero del catalogo: (77776-742)
Proveedor: Corning
Descripción: Optically clear PS. For applications where cell attachment is not desired.
Inquire for Price
El stock para este material es limitado pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
Este producto está marcado como restringido y sólo pueden ser adquiridos por cuentas aprobado para el envío. Si necesita más ayuda, contacte con nosotros en el (+52 55) 5005-0100 Opción 1.
Documentación adicional puede ser necesaria para comprar este artículo. Un representante de VWR se comunicará con usted si es necesario.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
Este producto ya no está disponible. Las alternativas pueden estar disponibles mediante la búsqueda con el número de catálogo VWR enumerado anteriormente. Si necesita más ayuda, por favor llame a Servicio al Cliente de VWR al (+52 55) 5005-0100 Opción 1.
1 - 16 of 31
no targeter for Bottom