Esta buscando: Termostatos con circulación
14  results were found
(IVA no incluido)
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
SearchResultCount:"14"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: VWR International
Descripción: VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40° to 200°C (depending on the controller).
Proveedor: VWR International
Descripción: VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40 to 200 °C (depending on the controller).
Product available on GSA Advantage®
Proveedor: VWR International
Descripción: VWR® Heated Circulating Baths offer precise temperature control, at temperature ranges from ambient +10° to 200°C (depending on the controller).
Product available on GSA Advantage®
Numero del catalogo: (89202-914)
Proveedor: VWR International
Descripción: VWR® Circulating Open Bath Systems include MX Temperature Controller and choice of stainless steel or polycarbonate reservoir.
Product available on GSA Advantage®
Numero del catalogo: (89202-886)
Proveedor: VWR International
Descripción: Create your own circulating bath system using your own vessel or a VWR® open bath tank.
Product available on GSA Advantage®
Numero del catalogo: (27476-014)
Proveedor: CARON PRODUCTS & SERVICES, INC. MX
Descripción: These recirculators acts as a water delivery system for use with any Caron humidity controlled chamber.
Product available on GSA Advantage® Small Business Enterprise
Numero del catalogo: (85022-312)
Proveedor: THERMO FISHER SCIENTIFIC MX
Descripción: This A10B refrigerated bath features a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from −10 to +100 °...
New Product
Numero del catalogo: (89421-862)
Proveedor: JULABO
Descripción: JULABO F Series chillers for basic laboratory cooling tasks. Ideal for replacing tap water. Provide recirculating fluid to applications at a consisten...
Water Efficient
Numero del catalogo: (10015-724)
Proveedor: JULABO
Descripción: CORIO™ heating circulators feature professional technology for routine laboratory tasks.
Numero del catalogo: (76477-128)
Proveedor: JULABO
Descripción: The MAGIO™ series of heating circulators operate with a working temperature range of 20 to +300 °C and heating capacity of up to 3 kW. With their powe...
Numero del catalogo: (89421-868)
Proveedor: JULABO
Descripción: JULABO recirculating coolers can handle virtually any cooling requirements in laboratories or industrial environments.
Energy Efficient
Water Efficient
Numero del catalogo: (71002-504)
Proveedor: POLYSCIENCE CORP-EQUIP/PARTS
Descripción: A common circulator anti-freeze fluid.
Numero del catalogo: (89202-370)
Proveedor: LAUDA-BRINKMANN INC
Descripción: ECO current replacement for Ecoline
Numero del catalogo: (71002-506)
Proveedor: POLYSCIENCE CORP-EQUIP/PARTS
Descripción: Propylene glycol is a safer alternative to ethylene glycol.
Product available on GSA Advantage®
Inquire for Price
El stock para este material es limitado pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
Este producto está marcado como restringido y sólo pueden ser adquiridos por cuentas aprobado para el envío. Si necesita más ayuda, contacte con nosotros en el (+52 55) 5005-0100 Opción 1.
Documentación adicional puede ser necesaria para comprar este artículo. Un representante de VWR se comunicará con usted si es necesario.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
Este producto ya no está disponible. Las alternativas pueden estar disponibles mediante la búsqueda con el número de catálogo VWR enumerado anteriormente. Si necesita más ayuda, por favor llame a Servicio al Cliente de VWR al (+52 55) 5005-0100 Opción 1.
1 - 14 of 14
no targeter for Bottom