Esta buscando: Muestreadores
61  results were found
(IVA no incluido)
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
SearchResultCount:"61"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: VWR International
Descripción: Dippers feature a 60 cm (235/8") long steel shaft encapsulated in PTFE for rigidity.
Proveedor: VWR International
Descripción: Samplers for thin to viscous liquids.
Numero del catalogo: (89026-192)
Proveedor: VWR International
Descripción: This PTFE molded sampling ladle is useful for transferring hot or cold reactive substances.
Proveedor: VWR International
Descripción: For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fi...
Numero del catalogo: (82024-578)
Proveedor: VWR International
Descripción: Designed for use in gas sampling or for pumping gas through apparatus.
Numero del catalogo: (10821-339)
Proveedor: BURKLE INC
Descripción: The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100.000 mPas.
Numero del catalogo: (75806-596)
Proveedor: BURKLE INC
Descripción: Sampler for deep-frozen and semi-solid substances products in the food industry.
Proveedor: COMERCIALOZADORA Y DISTRIBUIDORA MX
Descripción: The PowderDispo is a disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer samples can be take...
Numero del catalogo: (10821-261)
Proveedor: BURKLE INC
Descripción: Disposable zone samplers for taking samples from multiple layers of the sample material.
Proveedor: BURKLE INC
Descripción: These suction samplers are ideal for viscous substances such as pastes, mud, creams, oils.
Numero del catalogo: (10217-152)
Proveedor: BURKLE INC
Descripción: Specially developed for sample collection of aggressive liquids such as acids, alkalis, cleaning agents, (please check chemical resistance).
Numero del catalogo: (10217-062)
Proveedor: BURKLE INC
Descripción: Hand operated vacuum pump sampler, equipped with a 2.5 metre long PVC hose. Ideal for deep, narrow and hard to reach spots.
Proveedor: BURKLE INC
Descripción: Simple disposable sampler for taking samples of low viscosity media.
Proveedor: Bel-Art Products, a Part of SP
Descripción: This utensil’s specially curved shape permits contact with all corners of bottles and containers, virtually eliminating material or sample loss.
Proveedor: BURKLE INC
Descripción: The ergonomic handle of the DispoTube makes it easier to work and facilitates a good grip even under difficult conditions, for example with gloves.
Proveedor: Bel-Art Products, a Part of SP
Descripción: Designed for convenient sampling from tanks, vats, streams, ponds, lakes, etc., these economy dippers are lightweight, easy to use, and portable.
Inquire for Price
El stock para este material es limitado pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
Este producto está marcado como restringido y sólo pueden ser adquiridos por cuentas aprobado para el envío. Si necesita más ayuda, contacte con nosotros en el (+52 55) 5005-0100 Opción 1.
Documentación adicional puede ser necesaria para comprar este artículo. Un representante de VWR se comunicará con usted si es necesario.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
Este producto ya no está disponible. Las alternativas pueden estar disponibles mediante la búsqueda con el número de catálogo VWR enumerado anteriormente. Si necesita más ayuda, por favor llame a Servicio al Cliente de VWR al (+52 55) 5005-0100 Opción 1.
1 - 16 of 61
no targeter for Bottom