Esta buscando: Agitadores Vórtex
16  results were found
(IVA no incluido)
Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.
SearchResultCount:"16"
Sort Results
Vista lista Vista Extendida (Nueva)
Valore los resultados de su búsqueda
Proveedor: VWR International
Descripción: Fixed high speed mixing and variable speed models.
Numero del catalogo: (97043-562)
Proveedor: VWR International
Descripción: Variable speed analog mixer features a heavy-duty design and efficient motor that permits continuous duty operation and the ability to handle accessor...
Proveedor: VWR International
Descripción: These vortexers are designed to hold multiple test tubes, vials, or cylinders.
Numero del catalogo: (97043-564)
Proveedor: VWR International
Descripción: Ideal for applications that require repeatable results, the mixer features touchpad controls and LED displays for accurate speed and time results.
Product available on GSA Advantage®
Numero del catalogo: (89399-884)
Proveedor: VWR International
Descripción: The VWR® Advanced Microplate Vortex Mixer is ideal for applications that demand repeatable results.
Waste Reducing Product available on GSA Advantage®
Numero del catalogo: (89399-880)
Proveedor: VWR International
Descripción: The VWR® Standard Microplate Vortex Mixer is a variable speed analog mixer that is designed for continuous duty.
Waste Reducing Product available on GSA Advantage®
Numero del catalogo: (470230-486)
Proveedor: VWR International
Descripción: Powerful, Mini Vortex Mixer.
Numero del catalogo: (10153-688)
Proveedor: VWR International
Descripción: The Mini vortex mixer is the ideal vortex mixer for every lab with its compact size, ease of use, and its economic price point. The 3000 rpm fixed spe...
Numero del catalogo: (89028-150)
Proveedor: IKA WORKS, INC. MX
Descripción: MS3.4 MICROTITER PLATE ATTACHMENT FOR SHAKING MICROTITER PLATES
Numero del catalogo: (89209-860)
Proveedor: IKA WORKS, INC. MX
Descripción: Compact, universal small shaker suitable for shaking tasks with all small vessels and microtiter plates.
Numero del catalogo: (76417-524)
Proveedor: THERMO FISHER SCIENTIFIC MX
Descripción: Digital vortex mixer designed for accurate speed control when continuously mixing or pulsing samples.
Numero del catalogo: (89495-288)
Proveedor: IKA WORKS, INC. MX
Descripción: Designed for mixing small test samples, this new, attractively designed test tube shaker features touch operation and infinitely adjustable speed from...
Numero del catalogo: (89133-512)
Proveedor: Corning
Descripción: The Corning variable speed LSE Vortex Mixer provides fast, efficient mixing with minimal vibration
Low Manufacturing Impact
Proveedor: SCIENTIFIC IND
Descripción: Vortex Genie® Mixers are available in multiple models and with a vast array of available platforms and accessories.
Numero del catalogo: (10118-094)
Proveedor: ANTYLIA SCIENTIFIC
Descripción: The Vortamix Mini Vortexer from Argos Technologies is an unbelievably compact, yet powerful instrument engineered to mix tubes quickly and easily
Small Business Enterprise
Numero del catalogo: (89132-160)
Proveedor: ANTYLIA SCIENTIFIC
Descripción: Compact unit perfect for individual work stations.
Inquire for Price
El stock para este material es limitado pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
El stock para este material es limitada pero puede estar disponible en un almacén cerca de usted. Por favor, asegúrese de que ha iniciado sesión en la web para que el stock disponible se puede mostrar. Si el call sigue apareciendo y usted necesita ayuda, por favor llámenos al (+52 55) 5005-0123 o por email a miguel.sanchez@vwr.com
Este producto está marcado como restringido y sólo pueden ser adquiridos por cuentas aprobado para el envío. Si necesita más ayuda, contacte con nosotros en el (+52 55) 5005-0100 Opción 1.
Documentación adicional puede ser necesaria para comprar este artículo. Un representante de VWR se comunicará con usted si es necesario.
Este producto ha sido bloqueado por su organización. Por favor, pónganse en contacto con su departamento de compras para obtener más información.
El producto original ya no está disponible. Su sustituto se muestra a continuación.
Este producto ya no está disponible. Las alternativas pueden estar disponibles mediante la búsqueda con el número de catálogo VWR enumerado anteriormente. Si necesita más ayuda, por favor llame a Servicio al Cliente de VWR al (+52 55) 5005-0100 Opción 1.
1 - 16 of 16
no targeter for Bottom