Lauda® Aqualine Heated Water Baths, Brinkmann™
Proveedor: Lauda-Brinkmann
Lauda®
UE 029 UE 031 UE 036 LCZ 0689 UE 030 UE 032 UE 033 L000610
98000-106EA 45334.38 MXN
98000-106 98000-126 98000-136 98000-118 98000-120 98000-124 98000-112 98000-122
Lauda® Aqualine Heated Water Baths, Brinkmann™
Baños Baños de agua
Safe, durable water baths feature an integrated digital controller keypad with an LED that displays the temperature to 0.1°C
The tank is stainless steel with no welded seams and an exterior housing that features an electrostatic powder-paint finish and inner drip plates to prevent condensation. The internal chamber's corners are rounded for easy cleaning. A stainless steel hinged gable cover can be fixed in open position or removed completely to optimize bench space. The maximum temperature indicated can be reached only with the cover in use. Stainless steel racks secure vessels within bath, and have polymer coated handles for safe contact.
An embedded holder protects internal electronics, and a temperature cut-off function ensures samples will not be harmed if the temperature exceeds the desired point. The heating elements are restricted to the area below the bath vessel, ensuring homogenous temperature allocation and no exposed heating coils. The heating systems also make it possible to operate the bath down to a minimum water level of 20mm from the bottom of the tank, creating more space in the bath for large vessels
Operating temperature: 5...40 °C
Certificaciones: Baths are CE marked.
Información para pedidos: Model AL 12 can accommodate one RN 13, 18/1, 18/2, or 30 racks; any three RQ racks; or one platform. Model AL 18 can accommodate two RN 13, 18/1, 18/2, or 30 racks; any five RQ racks; or two platforms. Model AL 25 can accommodate two RN 13, 18/1, 18/2, or 30 racks; any five RQ racks; or two platforms. VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.