VWR® Reclosable Bags
Proveedor: VWR
VWR®
3620A 3735A 3730 3525A 3650A 3675A 3550A 3560A 3705A 3615A 3665A 3575A 3760 3685A 3700 3595A 3630A 3799 3765A 3795A 3695 3565A 3625A 3580A 3635A 3590A 3720A 3640A 3660A 3790 3660 3710A 3790A 3795 3670A 3720
89071-850CS 4690.18 MXN
89071-850 89073-086 89073-066 89073-088 89073-122 89073-100 89073-060 89073-082 89073-084 89073-062 89073-106 89073-128 89073-108 89073-046 89073-068 89073-102 89073-104 89071-798 89071-834 89071-812 89071-814 89073-090 89073-092 89073-132 89073-056 89073-072 89073-096 89073-074 89073-116 89071-848 89073-118 89073-134 89073-114 89071-822 89071-824 89071-846
VWR® Reclosable Bags
Bolsas Bolsas de almacenamiento
Reclosable bags are full gauge and offer exceptional clarity and quality.
  • LDPE resin fabrication
  • Reclosable design
  • Reinforced zipper locks in a smooth motion
Red line bags have a red line along the top of the bag and reinforced side-welded zipper to lock in a single smooth motion. Made of 100% virgin Low Density Polyethylene (LDPE) resin.
Certificaciones: Meet FDA/USDA specifications.