VWR® Stripe Autoclave Tape, Black
Proveedor: VWR
VWR®
VWR-T15-28-C
89097-924CS 234.95 MXN
89097-924
VWR® Stripe Autoclave Tape, Black
Indicadores de esterilización
Autoclave tape is used as a process indicator in steam sterilizers, and it is very well suited for the clinical and research environments.
  • Direct contact with substrates
Autoclave tape features an aggressive, pressure-sensitive adhesive designed to adhere to a variety of wraps so that the pack stays secure during sterilization and can be used on direct contact with substrates.
Tape also features a chemical indicator to verify that it has gone through the sterilization process. Black diagonal lines appear on the tape after 10 minutes at 121°C or 2 minutes at 134°C in a steam sterilizer.