VWR® Multi-Channel Ultra High-Performance Pipettors, Variable Volume
Proveedor: VWR, part of Avantor
P08111-VH200 P12111-VH200 PM555555VU P12111-VH010 P12555-VH050 P12111-VH300 P08555-VH200 P4908-50VWR PM111111VU P4908-200VWR P12555-VH300 P08555-VH010 P4908-300VWR P4908-10VWR P08555-VH050 P4912-300VWR P08111-VH010 P12555-VH200 P4912-50VWR P12111-VH050 P12555-VH010 P08111-VH300 P4912-10VWR P08111-VH050 P08555-VH300
89134-750EA 25416.7 MXN
10769-844 10769-812 10769-842 10769-820 10769-810 10769-840 10769-850 10769-808 10769-806 10769-818 10769-848 10769-838 10769-816 10769-846 10769-836 10769-814 89134-750 89134-746 89134-748 89134-758 89134-752 89134-744 89134-754
VWR® Multi-Channel Ultra High-Performance Pipettors, Variable Volume
VWR® Multi-Channel Ultra High-Performance Pipettors are equipped with a series of ergonomic enhancements not found on traditional pipettor models.
  • Ergonomic, comfortable, and extremely lightweight
  • Superior accuracy and precision
  • Four-digit volume dial for precision control
  • Universal tip fitting with adjustable tip ejector
Ultra high-performance pipettors feature an updated soft-touch plunger system that ensures surprisingly light aspiration and dispensing forces, while a levered ejection mechanism is designed to provide smooth and gentle tip ejection
The pipettor handle is constructed of a high-strength polypropylene polymer that combines excellent chemical resistance with thermal properties to permit autoclave sterilization. The handle permits both left- and right-handed use. A four-digit micrometer allows for fine control over volume adjustment and is located on the palm side of the device, facing the user at all times.
The volume adjustment wheel is conveniently located just beneath the plunger button and requires minimal force to select the desired volume. To prevent accidental volume changes, the adjustment wheel can be set to a locked position by pressing it downward.
Información para pedidos: Pipettors are supplied with a self calibration tool that, in conjunction with a balance, can be used for basic in-lab calibration. Additionally, all pipettors include color-coded identification tabs, a hanging shelf clip, and a calibration certificate.