Kanamicina sulfato USP
Peligro
VWRV0408-10G VWRV0408-25G VWRV0408-100G VWRV0408-1KG
97061-598EA 4414.23 MXN
97061-598 97061-600 75982-632 97061-602
Kanamicina sulfato USP
Kanamicin sulfato
An aminoglycoside antibiotic that inhibits protein synthesis in gram-negative and gram-positive bacteria and in mycoplasma. Recommended working concentration: 30µg/mL.
Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Ambiente
MDL Number: MFCD00070253
Núm. CAS: 25389-94-0
Order Now
Resultados test especificaciones
Potency (on anhydrous substance) Min. 750 μg/mg
Potency 0 - 9999 μg/mg
Appearance White to off-white powder
Identification A (IR) Passes test
Identification B (SO₄) Passes test
Identification C Passes test
Crystallinity Passes test
pH (25°C; 1 %) 6.5 - 8.5
Loss on drying Max. 4.0 %
Residue on ignition Max. 1.0 %
Chromatographic purity Passes test