Puromicina diclorhidrato, Ultra Pure Grade
Atención
VWRVJ593-25MG
97064-280EA 2189.6 MXN
97064-280
Puromicina diclorhidrato, Ultra Pure Grade
Puromycin dihydrochloride
A broad spectrum antibiotic that inhibits protein synthesis in both procaryotic and eucaryotic organisms
Widely used as a selection agent for cells carrying the Pac resistance gene encoding puromycin N-acetyl-transferase.
Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544.44 g/mol
MDL Number: MFCD00012691
Núm. CAS: 58-58-2