VWR® Premixed LB Broth
Proveedor: Avantor
VWR®
VWRVJ106-2KG VWRVJ106-10PK VWRVJ106-1KG VWRVJ106-500G
97064-110CS 22608.8 MXN
97064-110 97064-112 97064-114 97064-108
VWR® Premixed LB Broth
Medios Medios deshidratados para microbiología
Lysogeny broth (LB) is a nutritionally rich medium used for the growth of bacteria.
  • Save Time Using Premixed and Ready-to-Use Media
  • Homogenous and High Quality Powders
Lysogeny broth (LB) is a nutritionally rich medium used for the growth of bacteria. LB continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli. Powdered LB is available in premixed Miller or Lennox formulations and in Ready-Pack™ pouches which prepare 1L of media.