VWR® Life Science HistoChoice® Mounting Media
Proveedor: Avantor
HistoChoice®
VWRVH157-120ML VWRVH157-475ML
97060-990EA 2903.83 MXN
97060-990 97060-988
VWR® Life Science HistoChoice® Mounting Media
Tintes Reactivos de microscopía y tinción
HistoChoice® Mounting Media was designed for use with HistoChoice® Clearing Agent, but can also be used as a separate stand-alone medium for standard slide preparation. It is suitable for both thick or thin specimen mounting. It can also be used with immersion oil. The low viscosity of this mounting media means easier application, resulting in bubble-free preparations that save valuable lab time.
  • Does not crack or splinter with age
  • Will not discolor
  • Low viscosity for bubble-free slide preparations
  • Works for all types/sizes of specimens