VWR® Acryl-Glide Biotechnology grade
Proveedor: Avantor
VWR®
VWRVE319-100ML
97064-746EA 480.61 MXN
97064-746
VWR® Acryl-Glide Biotechnology grade
Reactivos para electroforesis Western Blotting Reagents
Acryl-Glide Solution and Wipes are designed for electrophoresis plate coating and provides a hydrophobic layer that outperforms even the best silane coatings. With acryl-glide, coated plates can be reused several times with a single application and still allow for smooth and complete transfer of gels without breaking or gel/plate adhesion.
This product has been functionally tested on denaturing and non-denaturing polyacrylamide gels and has no effect on migration distance or resolution of nucleic acids.