Penicilina G sal sódica Y-IRR, en polvo, irradiado

Proveedor: VWR CHEMICALS LLC - SANBORN

VWRVE480-20ML
97062-798EA 761.55 MXN
97062-798
Penicilina G sal sódica Y-IRR, en polvo, irradiado
[2S-(2α,5α,6β)]-6-(Fenilacetamido)-3,3-dimetil-7-oxo-4-tia-1-azabiciclo[3.2.0]heptano-2-carboxilato de sodio

These antibiotic and antimycotic mixtures have been aseptically prepared from the highest quality materials and are gamma-irradiated to ensure sterility. The dry blends, or lyophilized powders, are packaged in amber serum vials with multiple injection stoppers for syringe mediated liquid handling. Simply reconstitute the powder in the recommended volume of sterile tissue culture grade water.


Each reagent has been tested for compatibility with standard tissue culture systems and remains stable for greater than one year when stored as recommended. All powders and solutions should be stored at 4°C unless otherwise noted. Aseptically prepared and gamma irradiated to ensure sterility. Functionally tested for compatibility with standard tissue culture systems. When reconstituted in 20 mL sterile water, contains 10,000 U/ml Penicillin G.Inhibits cell wall synthesis in gram-positive and gram-negative bacteria

Formula: C₁₆H₁₇N₂NaO₄S
MW: 356.38 g/mol
Mezcla Pt: 255 °C
Densidad: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambiente
MDL Number: MFCD00069666
Núm. CAS: 69-57-8
EINECS: 200-710-2

Order Now

Resultados test especificaciones

pH (6% water at 25°C) 5.0 - 7.5
Potency (Anhydrous) 1500 - 1750 u/mg

Algunos números de catálogo podrían haber cambiado derivado de una estandarización global. Los productos o empaques no están afectados.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR