VWR® Bis-2™, 2% Solution of Bis-Acrylamide
BIS-2™
VWRVM193-500ML
97063-948EA 1285.2 MXN
97063-948
VWR® Bis-2™, 2% Solution of Bis-Acrylamide
  • Ultra-pure powders with acrylamide purity > 99.9%
  • Convenient ready-to-use
  • Eliminates hazardous powdered acrylamide handling
VWR offers an extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions, and ready-to-use solutions for customized polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids. Our ultra pure (> 99.9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimize the handling of neurotoxic acrylamide.