VWR® EZ-Vision® In-Gel Solution, 10000X
Proveedor: Avantor
VWR® EZ-Vision®
VWRVN391-0.5ML VWRVN391-5MLDRP VWRVN391-15MLDRP
97064-174EA 3046.4 MXN
97064-174 97068-178 97068-180
VWR® EZ-Vision® In-Gel Solution, 10000X
Tintes Colorantes para electroforesis
  • Immediate results
  • Easy and convenient
  • Eliminates the use of ethidium bromide
In-Gel Solution, 10,000X is a non-mutagenic, non-toxic fluorescent DNA Dye that is used as an in-gel or post-run stain to visualize DNA in agarose gels. EZ-Vision® In-Gel Solution is an excellent replacement for the often used, but hazardous ethidium bromide.  As an in-gel preparation for electrophoresis, EZ-Vision® In-Gel Solution is simply added to molten agarose prior to gel casting, allowing instant visualization of DNA bands upon UV exposure. Alternatively, EZ-Vision® In-Gel Solution can be used to stain the gel after electrophoresis.