VWR® Agarose RA™ Agarose RA
Proveedor: Avantor
Agarose RA™
VWRVN605-500G VWRVN605-100G VWRVN605-250G VWRVN605-1KG
97064-250CS 73863.82 MXN
97064-250 97064-252 97064-254 97064-258
VWR® Agarose RA™ Agarose RA
Geles de eletroforesis Agarosa y acrilamida
A quality general application agarose that provides good resolution and is cost-effective for high volume users.
  • Ideal Agarose for High Throughput
  • High Resolution
  • Easy Preparation
  • Excellent Sensitivity
A quality general application agarose that provides good resolution and is cost-effective for high volume users. It was developed specifically for quick checks of PCR products, plasmids preps, screening and cloning. Features high resolution, easy preparation and set up, a low EEO and excellent sensitivity with low background when stained.