VWR® RiboZol™ RNA Extraction Reagent
Proveedor: Avantor
RiboZol™
VWRVN580-200ML VWRVN580-30ML
97064-950CS 707.19 MXN
97064-950 97064-952
VWR® RiboZol™ RNA Extraction Reagent
Tampones Tampones de extracción
This extraction reagent is a ready-to-use, single phase phenol solution for the isolation of total RNA from even the most difficult cell and tissue types
  • Isolate intact, undegraded total RNA from even the most difficult of cells and tissues types
  • Extract total RNA in less than 1 hour
  • Achieve consistent results and excellent recovery of small RNAs
It is optimized to produce high quality RNA from a variety of biological samples. The sample is lysed directly in the reagent to inhibit RNase activity and prevent RNA degradation during purification. RNA obtained by the RiboZol™ purification procedure is compatible with a variety of downstream applications.