VWR® Next Gel® Gel Electrophoresis Solutions, Polyacrylamide
Proveedor: Avantor
NEXT GEL®
VWRVM256-500ML VWRVM258-500ML
97063-026EA 3736.72 MXN
97063-034 97063-026
VWR® Next Gel® Gel Electrophoresis Solutions, Polyacrylamide
Geles de eletroforesis Agarosa y acrilamida
NEXT GEL® is a ready to pour pre-mixed solution of acrylamide, bisacrylamide, gel buffer, and SDS that enables superior, ultra-fine resolution of protein bands. The unique acrylamide matrix slows the migration of proteins, eliminating the need for a stacking gel. This not only saves time, but also permits the proteins to run across a longer gel surface resulting in increased resolution. The proprietary chemistry has gradient-like properties, which enables the separation of small peptides and high molecular weight proteins in the same gel.
  • Ready to pour SDS polyacrylamide solutions - just add APS and TEMED
  • Faster gel casting with no stacking gel required
  • Broad range of separation - 3,5 kDa to 212 kDa on the same gel
  • High resolution of protein bands
Información para pedidos: Supplied with 20X running buffer.