Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Proveedor: Simport
HISTOSETTE®
M492-10 M492-3 M492-4 M492-8 M492-8 M492-16 M492-3 M492-4 M492-16 M492-17 M492-11 M492-7 M492-12 M492-2 M492-11 M492-10 M492-6 M492-9 M492-2 M492-9 M492-5 M492-5 M492-7 M492-18 M492-6
60872-524CS 5168.39 MXN
CA73960-072 CA73960-070 CA73960-054 60872-520 CA73960-056 60872-536 87002-452 60872-532 60872-534 CA11031-920 87002-454 60872-518 10040-366 CA73960-060 CA73960-064 CA73960-062 60872-530 CA73960-068 CA73960-066 CA73960-512 60872-526 60872-528 60872-522 CA73960-514 60872-524
Histosette® II Tissue Processing/Embedding Cassettes with Integrated Lids, Simport
Casetes para histología Casetes para procesamiento de tejidos
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage
Molded from a special high-density polymer, they can keep specimens safely submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids can be opened and closed as often as necessary and will always relock securely without danger of specimen loss. The unique front hinge design allows for single-hand cassette manipulation. The anterior writing area is slanted at a 45° angle to make the cassette more suitable with certain types of cassette labeling instruments
Información para pedidos: Packed in three dispenser packs of 500 cassettes.