ClipTip technology locks tips firmly in place and ensures a complete seal on every channel with minimal tip attachment and ejection forces using Thermo Scientific™ F1-ClipTip™ Multichannel pipettes.
- Higher-quality results
- More efficient research
- Remains sealed until ejection
- Lightweight design
- Reproducible from user to user
- Secure volume setting
- Lifetime manufacturer warranty
Newly enhanced volume adjustment allows for a silent, effortless and ergonomic user experience. Eliminate the frustration of banging tips on your pipette. With a light touch, each tip is clipped securely on the pipette. Reduce the risk of loose or leaking tips. When ClipTip pipette tips are on, they are sealed and will not loosen.
The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles. Enable consistent and reproducible pipetting from user to user.
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase in air boost ensures efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.
Certificaciones: The F1-ClipTip Pipette together with compatible ClipTip pipette tips is CE marked in accordance with IVD Directive in Europe.
Información accesorios: Wide selection of ClipTip pipette tips available. Choose from five sizes in 96-format: 20, 50, 200, 300, and 1000 μl.
Información para pedidos: Industry-leading lifetime warranty with convenient online registration; may vary by country.