PYREX® Desiccators with 24/29 Standard Taper Stopcock, Vacuum Rated, Corning
Proveedor: Corning
PYREX®
3121-200 3121-STPK 3121-200CO 3121-150 3121-250 3121-150CO
89134-408CS 7500 MXN
89134-410 89134-408 89134-404 89134-406 89134-414 89134-402
PYREX® Desiccators with 24/29 Standard Taper Stopcock, Vacuum Rated, Corning
Desecadores Desecadores de vidrio
These PYREX® desiccators have a rugged 24/29 standard tapered stopcock with a tooled hose connection that serves as the vacuum inlet.
  • The tubulation Ext.Ø is approximately 10 mm
  • Material is Borosilicate Glass Code 7740
  • This design eliminates the need for grease
It is designed so that its manipulation is not affected by the vacuum. To introduce vacuum into the desiccator, loosen the stopcock to disengage the PTFE plug from contact with the glass. A half turn from this position closes the desiccator.
Replacement covers and stopcock are listed in the Order tab below. The recommended replacement lower bowls which are interchangeable with these desiccators, are as follows: for the 2.4 L desiccator, use replacement bowl Cat. No. 89134-388; for the 5.8 L desiccator, use replacement bowl Cat. No. 89134-394; and for the 10.5 L desiccator, use replacement bowl Cat. No. 89134-400.
Información para pedidos: Plate is not included.
Precaución: Warning: Do not heat or subject to pressure. Rated to 1-atmosphere vacuum.