Accu-Safe Enclosed Chamber Bottle Thermometers, Thermco
Proveedor: Thermco
ACCR20001S ACCR0201S ACCOV080S ACCF100S ACCDB301 ACCOV700S ACCI310S ACCI0301S ACCBBS ACCF0101S ACCULF010S ACCAM0401S ACCOV070S ACCBB210S ACCI300S ACCDB40
89137-076EA 1326.91 MXN
89137-082 10145-502 89137-084 10145-556 10145-500 89137-086 89137-076 10145-498 89137-088 10145-496 89137-078 10145-550 10145-494 10145-492 10145-490 89137-080
Accu-Safe Enclosed Chamber Bottle Thermometers, Thermco
Termómetros Analogue Thermometers
These spirit filled, non-mercury thermometers are housed in unbreakable plastic bottles filled with a non-toxic insulating liquid, used to ensure accurate temperature readings without being affected by the opening and closing of a chamber door.
Thermometers are fluoropolymer coated for additional protection. All thermometers have thoroughly annealed glass and engraved stems for lasting accuracy in 0.1, 0.5 and 1 °C divisions. Magnets are included to attach thermometers to a steel door or wall.
Información para pedidos: All thermometers have their own unique serial number and are supplied with a NIST traceable statement.