Sterile Syringe Convenience Trays, BD Medical
Proveedor: BD Medical
305618 305617 309680 309701 309680 305617 309702 309701 305618 309703 309605 309702 309605
BD309702CS 140 MXN
BD309702 BD309701 CABD309680 CABD309701 CABD309702 CABD309605 BD309680 CA-BD305617 CA-BD305618 BD309605 BD309703 BD305618 BD305617
Sterile Syringe Convenience Trays, BD Medical
Jeringas Jeringas médicas
These convenient trays provide sterile syringes with fully depressed stoppers in a single-direction orientation to assist in batch-preparation of syringes
The stopper is positioned at the bottom-most part of the tray to reduce manipulation