NORM-JECT® Catheter Sterile Syringe, Air-Tite Products
Proveedor: Air-Tite
NORM-JECT®
4850002000
53548-015CS 10946.63 MXN
53548-015
NORM-JECT® Catheter Sterile Syringe, Air-Tite Products
Jeringas Jeringas médicas
NORM-JECT® syringes feature medical-grade plastics, including a polypropylene barrel and a polyethylene plunger. Great value at reasonable costs.
  • Sterile
  • Certified DNA free
  • Latex free
  • Rubber free
  • Silicone free
  • Styrene free
  • DEHP free
These syringes are the choice for any situation needing an inert, non-reactive syringe. Because of their composition, these syringes are indicated for Nuclear Medicine, Amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures. They are more chemically resistant than rubber tipped syringes. These inert syringes do not contain contaminates often associated with disposables.
The syringes can be used for sampling, mixing, and filtering. They are smooth drawing with a positive safety stop to prevent accidental spills. They feature accurate, easy-to-read increments. Their dose saver design minimizes waste, saving up to 4.5 ml of medication per 100 syringes used, as compared to non low deadspace Luer syringes (Actual savings may vary).
Certificaciones: Meets ISO Standards, CE Registered, FDA Registered.
Información para pedidos: Sterile and individually wrapped.