Corning® Gosselin™ Inoculating Loops
Proveedor: Corning
Corning®
OS1B-01 OS10B-02 OS10-02 OS1-01
89200-928 89200-922 89200-932 89200-924
Corning® Gosselin™ Inoculating Loops
Esparcidores Asas de siembra
These polystyrene inoculating loops are highly ergonomic and are color coded for easy identification.
  • Spherical or needle extremity
  • Assured sterile 10-3
Traceability information is provided on each bag.