SMITH & WESSON® CALIBER™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Proveedor: Kimberly-Clark
Smith & Wesson®
23006 23008 23010 23007
89130-954EA 378.87 MXN
89130-958 89130-954 89130-956 89130-952
SMITH & WESSON® CALIBER™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Gafas
Ergonomic, rubber nose pads with built-in airflow holes will adjust to fit the shape of any nose for an ideal fit.
  • Contoured, soft-touch rubber temples improve wearer comfort
  • Base curve 7
  • Scratch-resistant polycarbonate lens
  • 99.9% UV protection
Cut-to-shape lens provides less distortion around the edges, and a hinge stop helps prevent lens damage.
Certificaciones: ANSI Z87.1