Nalgene® Polysulfone Reusable Bottle Top Filters, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
DS0320-2545 DS0320-5045 DS0320-5033
28199-325EA 6404.58 MXN
28199-325 28199-322 28199-324
Nalgene® Polysulfone Reusable Bottle Top Filters, Thermo Scientific
Filtros Filtros de la parte superior de botellas
Nalgene® Polysulfone Reusable Bottle Top Filters offer the convenience of a bottle top filter with the cost savings and waste reduction of a reusable filter housing.
The filters feature a removable sterilization membrane support plate designed to provide maximum flow rate and throughput, as well as a vacuum port on the lower half of the assembly and molded-in graduations. The polypropylene tubing adapter can be stuffed with cotton for sterile venting during autoclaving.
The deep threads screw securely onto the bottle for convenient filling and vacuum filtration.