VWR® Protective Eyewear with Wraparound Lens
Proveedor: VWR
VWR®
PP-01BLUE 5330 PP-01MIRROR PP-01SMOKE
89187-986CS 5830.07 MXN
89187-986 76329-626 76329-624 76329-622
VWR® Protective Eyewear with Wraparound Lens
Gafas
VWR® protective eyewear with wraparound lens offers stylish and secure personal protection, with an array of comfort features to assist the wearer throughout extended use.
  • Fit comfortably with half-mask respirators
  • Impact-resistant, scratch-resistant polycarbonate lens
  • Provides 99.9% UV protection
Super lightweight wraparound eyewear for men and women combines a sleek appearance with rugged durability. Eyewear features rubberized temple tips and an integrated nose piece for extra comfort.
Certificaciones: ANSI Z87.1 certified.