Allegra® X-14 Series Benchtop Centrifuges 120 V, Beckman Coulter
Proveedor: Beckman Coulter
Allegra®
B08882 B08860 B08884 B08874 B08862 A99464 B08872 B08894 B08882 B08861 B08862 B08894 B08884 B08896 B08872 B08874 A99464 B08896 B08860
CABKB08894 CABKB08861 CABKB08872 CABKB08862 CABKB08884 CABKB08896 CABKB08874 BKB08874 BKB08896 BKB08884 BKB08862 BKB08872 BKB08894 BKB08860 BKB08882 BKA99464 CABKB08882 CABKB08860 CABKA99464
Allegra® X-14 Series Benchtop Centrifuges 120 V, Beckman Coulter
Centrífugas Centrífugas de sobremesa
The Allegra® X-14 Series accommodates the SX4750A ARIES self-balancing swinging bucket rotor, the SX4750 swinging bucket rotor, and the FX6100 fixed angle rotor.
  • ARIES Smart Balance Rotor (SX4750A) Automatically Detects and Corrects Imbalance up to 50 g
  • 4 x 750 mL Swinging Bucket Rotor (up to 4 300 x g/4300 rpm)
  • Allegra X-14R (Refrigerated) Achieves 4°C in Less Than Four Minutes
  • 3 L Maximum Capacity
  • Extensive Biocontainment Options
Cell Culture Packages also include 15 mL (set of four) and 50 mL (set of four) conical adapters
Blood Sample Preparation Application Packages also include 13 mm (set of four) and 16 mm (set of four) diameter tubes. Maximum capacity is 120x13 mm and 76x16 mm blood tubes per run. BioSafe Aerosolve Canister Packages also include a set of four Aerosolve canisters, four 15 mL conical tube racks, and four 50 mL conical tube racks. Maximum rack capacities are 24x15 mL and 16x50 mL. Microplate Application Packages hold up to sixteen standard microplates or four deep well plates.
Maximum capacity for these adapters is 56x15 mL, and 28x50 mL.