Going beyond protection and compliance, AccuFlow™ Type II Safety Cans provide fast, precision pouring when handling flammable liquids. Patented, innovative manifold automatically vents for smooth, glug-free liquid flow, while the comfortable Safe-Squeeze® trigger offers maximum control when pouring. A 9" (229mm) flexible metal hose offers targeted pouring into small openings. Quick-fill port with ergonomically designed lever handle makes filling easier than ever. Protection features include a long lasting, stainless steel flame arrester that dissipates heat to prevent flashback ignition and a gasketed, self-closing leakproof that automatically vents to guard against rupture or explosion. Warning label with extra large ID zone permits user to identify contents, department location, or individual user name. All can sizes have extra capacity for mixing in additives for two cycle engines.
- Built-in flexible metal hose allows smooth, accurate, spill-free pouring into small openings
- All cans have a flame arrester and are tested under pressure to guarantee leakproof construction
- Heavy-duty steel with red powder-coat finish protects against abrasion and rough use
- FM approved and C-UL listed
Certificaciones: FM. UL, ULC, TUV.