sensION+ Benchtop Meter Bundles, Hach
Proveedor: Hach
LPV3110.97.0002 LPV3170.97.0002 LPV2010T.97.002 LPV2100.97.0002 LPV2121T.97.002 LPV2210T.97.002 LPV2014T.97.002 LPV2200.97.0002 LPV2021T.97.002 LPV2111T.97.002 LPV2200.97.0002 LPV4140.97.0002 LPV3010.97.0002 LPV2221T.97.002 LPV2011T.97.002 LPV2214T.97.002 LPV4110.97.0002 LPV2211T.97.002 LPV3070.97.0002 LPV2000.97.0002 LPV2114T.97.002 LPV2110T.97.002 LPV2114T.97.002 LPV2211T.97.002
89174-142EA 77584.4 MXN
CA11028-130 89174-122 89174-144 89174-176 89174-132 89174-124 89174-146 89174-134 89174-178 89174-148 89174-126 CA89174-146 89174-136 89174-128 89174-138 89174-180 89174-182 89174-140 89174-150 89174-172 89174-142 89174-174 89174-130 CA11028-126
sensION+ Benchtop Meter Bundles, Hach
Medidores electroquímicos Medidores de pH
Hach sensION+, an all-in-one system with guided menu navigation makes general electrochemistry testing fast and simple. Each system is designed to be used in a wide variety of applications and comes complete with everything you need to start testing.
A guided navigation menu allows you to follow simple prompts to set up your measurements, thus reducing the time necessary to start testing and improving the accuracy of measurements.
All benchtop kits come complete with meter, probe, integrated magnetic stirrer, electrode stand, standards, calibration flasks and all required accessories.
The extensive number of probes built for sensION+ means you have your choice of probes for every testing environment. Several calibration modes are provided with automatic recognition of multiple standards.
Certificaciones: CE marked. WEEE compliant.