sensION+ Portable Meter Bundles, Hach
Proveedor: Hach
LPV4000970002 LPV2555970002 LPV3500970002 LPV2645970002 LPV2500970002 LPV2550T97002 LPV3560970002 LPV3562970002 LPV4048970002 LPV2552T97002
89174-164EA 30732.65 MXN
89174-160 89174-192 89174-170 89174-162 89174-184 89174-164 89174-186 89174-152 89174-154 89174-156
sensION+ Portable Meter Bundles, Hach
Medidores electroquímicos Medidores de pH
The nnovative menu designs and simple keypads result in truly intuitive operation for the sensION+ portable meter.
The sensION+ portable meter makes simple and fast measurements, has temperature readjustment capability and has an ergonomic shape that allows for one hand operation. The extensive number of probes built for sensION+ means that there is a choice of probes for nearly every testing environment.
A guided menu allows you to follow simple prompts to set up the measurements, thus reducing the time necessary to start testing and improving the accuracy of measurements.
Certificaciones: CE.
Información para pedidos: Available as a kit or meter only.