sensION+ pH Electrodes, Hach
Proveedor: Hach
E16M323 LZW5050T.97.002 E16M400 E16M329 E16M301 E16M400 LZW5052T.97.002 E16M343 E16M320 B10C161 E16M313 E16M318 E11M001 E16M319 LZW5010T.97.002 LZW5011T.97.002 LZW5014T.97.002 LZW5014T.97.002 LZW5051T.97.002 LZW5050T.97.002 B10B250 E16M329 E16M500 E11M004 LZW5021T.97.002 E16M303 E16M315 E16M334
89174-198EA 20200 MXN
89174-218 CA89174-248 89174-254 89174-298 89174-198 89501-026 89174-256 89174-278 89174-258 89174-216 89174-250 89174-194 89174-196 CA11028-178 CA89501-026 89174-264 89174-220 89174-286 89174-242 89174-300 CA89174-198 89174-200 89174-244 89174-302 89174-268 89174-246 89174-248 89174-284
sensION+ pH Electrodes, Hach
Electrodos Electrodos pH
To meet your analysis needs, combined pH electrodes are available with a variety of lengths, diameters and reference systems.
Our sensION+ high performance pH electrodes have encapsulated reference element, Ag⁺ ion barrier (except 5010T) and are combination type including built in Pt 1000 temperature sensor.
pH glass electrodes from Radiometer Analytical are made with highly shock-resistant glass, making them extremely tough. All pH probes may be used with sensION+ meters.
Certificaciones: CE marked. WEEE compliant.