Reference Electrodes, Hach
Proveedor: Hach
B20C820 B20B300 LZW5044.97.0002 E21M003
89174-314EA 16900 MXN
89174-316 89174-228 89174-312 89174-314
Reference Electrodes, Hach
Electrodos Electrodos pH
A reference electrode provides a stable potential whatever the measurement conditions.
These reference electrodes have a glass body and are available with a variety of lengths, diameters and reference systems to meet your analysis needs. The distinguishing characteristics between reference electrodes are the type of reference system and the liquid junction.
Certificaciones: CE.WEEE.