Conductivity Electrodes, Hach
Proveedor: Hach
E61M013 E61M008 LZW5070.97.0002 E61M011 LZW5062.97.0002 LZW5060.97.0002 E61M022
89174-206EA 22850.02 MXN
89174-206 89174-360 89174-370 89174-362 89174-222 89174-368 89174-224
Conductivity Electrodes, Hach
Electrodos Sensores de conductividad
Hach offers conductivity cells for a wide variety of applications
It is important to choose a cell with the right construction and geometry for your particular application and working conditions.