Cryogenic Storage Labels, Argos Technologies
Proveedor: Argos Technologies
PolarSafe™
04396-86 04396-81 04396-69 04396-89 04396-79 04396-91 04396-76 04396-87 04396-75 04396-85 04396-64 04396-66 04396-73 04396-90 04396-71 04396-88 04396-83 04396-80 04396-68 04396-65 04396-74 04396-77 04396-82 04396-67
89207-076RL 1735.16 MXN
89207-084 89207-094 89207-082 89207-092 89207-080 89207-090 89207-102 10030-116 89207-112 89207-078 89207-100 89207-110 89207-088 89207-076 89207-098 89207-086 89207-074 89207-108 30618-663 30618-661 89207-104 30618-667 30618-665 30618-669
Cryogenic Storage Labels, Argos Technologies
Etiquetas
Argos PolarSafe™ dots and labels are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plastic ware.
  • Label holder temperature range: –60 to 70 °C
These unique labels will not only withstand conventional freezing (–80 °C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at –196 °C). No need for separate labeling solutions for your cryo-preservation units and your ultra low freezers. The labels can even be used in incubators.